Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Nitroindazole, Tocris Bioscience™

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
2942-42-9
C7H5N3O2
163.136
PQCAUHUKTBHUSA-UHFFFAOYSA-N
7-nitroindazole,1h-indazole, 7-nitro,7-ni,7-nitro-indazole,unii-ux0n37cmvh,ccris 3309,2h-indazole,7-nitro,ux0n37cmvh,7ni
1893
7-nitro-1H-indazole
C1=CC2=C(C(=C1)[N+](=O)[O-])NN=C2
This item is not returnable. View return policy
2942-42-9
C7H5N3O2
163.136
PQCAUHUKTBHUSA-UHFFFAOYSA-N
7-nitroindazole,1h-indazole, 7-nitro,7-ni,7-nitro-indazole,unii-ux0n37cmvh,ccris 3309,2h-indazole,7-nitro,ux0n37cmvh,7ni
1893
7-nitro-1H-indazole
C1=CC2=C(C(=C1)[N+](=O)[O-])NN=C2

Non-selective NOS inhibitor

Reversible, competitive and non-selective NOS inhibitor.

Monosodium Salt also available.

Quantity 50mg
Formula Weight 163.14
Percent Purity >98%
Chemical Name or Material 7-Nitroindazole

Recommended Storage : Desiccate at -20°C