Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Nitroindole-3-carboxaldehyde, 97%

Click to view available options
Quantity:
250mg
1g
5g
10553-14-7
C9H6N2O3
190.158
MFCD01074519
ADGKBVRTGVODMM-UHFFFAOYSA-N
7-nitroindole-3-carboxyaldehyde,7-nitroindole-3-carboxaldehyde,7-nitroindole-3-carbaldehyde,3-carboxy-7-nitro-1h-indole,7-nitro-1h-indole-3-carboxaldehyde,7-nitroindole-3-aldehyde,pubchem9125,3-formyl-7-nitro-1h-indole,7-nitroindole 3-carboxaldehyde
2762438
7-nitro-1H-indole-3-carbaldehyde
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2C=O
10553-14-7
C9H6N2O3
190.158
MFCD01074519
ADGKBVRTGVODMM-UHFFFAOYSA-N
7-nitroindole-3-carboxyaldehyde,7-nitroindole-3-carboxaldehyde,7-nitroindole-3-carbaldehyde,3-carboxy-7-nitro-1h-indole,7-nitro-1h-indole-3-carboxaldehyde,7-nitroindole-3-aldehyde,pubchem9125,3-formyl-7-nitro-1h-indole,7-nitroindole 3-carboxaldehyde
2762438
7-nitro-1H-indole-3-carbaldehyde
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2C=O
Quantity 250mg
Formula Weight 190.16
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 7-Nitroindole-3-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold