Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Nitroindole, 97%

Click to view available options
Quantity:
1g
5g
25g
6960-42-5
C8H6N2O2
162.148
MFCD00005683
LZJGQIVWUKFTRD-UHFFFAOYSA-N
7-nitroindole,1h-indole, 7-nitro,indole, 7-nitro,7-nitro indole,pubchem7441,acmc-1b2jq,5-20-07-00044 beilstein handbook reference,ksc353k7l,1h-indole, 7-nitro-9ci
23396
7-nitro-1H-indole
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2
6960-42-5
C8H6N2O2
162.148
MFCD00005683
LZJGQIVWUKFTRD-UHFFFAOYSA-N
7-nitroindole,1h-indole, 7-nitro,indole, 7-nitro,7-nitro indole,pubchem7441,acmc-1b2jq,5-20-07-00044 beilstein handbook reference,ksc353k7l,1h-indole, 7-nitro-9ci
23396
7-nitro-1H-indole
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2
Quantity 5g
Melting Point 94°C to 96°C
Odor Odorless
Beilstein 130874
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 7-Nitroindole

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P281-P308+P313-P501c

H341

EINECSNumber : 000-000-0

RTECSNumber : NM1169000

TSCA : No

Recommended Storage : Ambient temperatures