Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Octenyl Acetate 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
5048-35-1
C10H18O2
170.252
MFCD01861278
DBRVFNFZYZFKRC-UHFFFAOYSA-N
8-Acetoxy-1-octene, Acetic Acid 7-Octenyl Ester
13990525
oct-7-enyl acetate
CC(=O)OCCCCCCC=C
5048-35-1
C10H18O2
170.252
MFCD01861278
DBRVFNFZYZFKRC-UHFFFAOYSA-N
8-Acetoxy-1-octene, Acetic Acid 7-Octenyl Ester
13990525
oct-7-enyl acetate
CC(=O)OCCCCCCC=C
Boiling Point 104°C
Color Colorless
Quantity 25g
Formula Weight 170.25
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 7-Octenyl Acetate

EINECSNumber : (2)-0760

TSCA : No