Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Oxabicyclo[2.2.1]heptane, 98%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
279-49-2
C6H10O
98.145
MFCD00082342
YPWFNLSXQIGJCK-UHFFFAOYSA-N
1,4-epoxycyclohexane,7-oxabicyclo 2.2.1 heptane,7-oxanorbornane,1,4-oxycyclohexane,3,6-endoxocyclohexane,3,6-endooxycyclohexane,cyclohexane,4-epoxy,cyclohexane,6-endoxo,cyclohexane, 1,4-epoxy
9234
7-oxabicyclo[2.2.1]heptane
C1CC2CCC1O2
279-49-2
C6H10O
98.145
MFCD00082342
YPWFNLSXQIGJCK-UHFFFAOYSA-N
1,4-epoxycyclohexane,7-oxabicyclo 2.2.1 heptane,7-oxanorbornane,1,4-oxycyclohexane,3,6-endoxocyclohexane,3,6-endooxycyclohexane,cyclohexane,4-epoxy,cyclohexane,6-endoxo,cyclohexane, 1,4-epoxy
9234
7-oxabicyclo[2.2.1]heptane
C1CC2CCC1O2
Flash Point 12°C (55°F)
Quantity 25g
Boiling Point 118°C to 120°C (713mmHg)
UN Number UN3271
Formula Weight 98.15
Percent Purity 98%
Chemical Name or Material 7-Oxabicyclo[2.2.1]heptane

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.