Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Oxabicyclo[2.2.1]heptane 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
279-49-2
C6H10O
98.145
MFCD00082342
YPWFNLSXQIGJCK-UHFFFAOYSA-N
1,4-epoxycyclohexane,7-oxabicyclo 2.2.1 heptane,7-oxanorbornane,1,4-oxycyclohexane,3,6-endoxocyclohexane,3,6-endooxycyclohexane,cyclohexane,4-epoxy,cyclohexane,6-endoxo,cyclohexane, 1,4-epoxy
9234
7-oxabicyclo[2.2.1]heptane
C1CC2CCC1O2
279-49-2
C6H10O
98.145
MFCD00082342
YPWFNLSXQIGJCK-UHFFFAOYSA-N
1,4-epoxycyclohexane,7-oxabicyclo 2.2.1 heptane,7-oxanorbornane,1,4-oxycyclohexane,3,6-endoxocyclohexane,3,6-endooxycyclohexane,cyclohexane,4-epoxy,cyclohexane,6-endoxo,cyclohexane, 1,4-epoxy
9234
7-oxabicyclo[2.2.1]heptane
C1CC2CCC1O2
Color Colorless
Refractive Index 1.4470 to 1.4490
Packaging Glass bottle
Boiling Point 119.0°C (713.0 mmHg)
Quantity 5g
Flash Point 12°C
Density 0.9600g/mL
Infrared Spectrum Authentic
Specific Gravity 0.96
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 98.14
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 7-Oxabicyclo[2.2.1]heptane, 99%
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger