Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Pentadecanone, 98%

Click to view available options
Quantity:
1g
5g
25g
6064-38-6
C15H30O
226.404
MFCD00027335
VUXXXYMMZBAUSE-UHFFFAOYSA-N
7-pentadecanone,n-hexyl n-octyl ketone,acmc-1b06x
292625
pentadecan-7-one
CCCCCCCCC(=O)CCCCCC
6064-38-6
C15H30O
226.404
MFCD00027335
VUXXXYMMZBAUSE-UHFFFAOYSA-N
7-pentadecanone,n-hexyl n-octyl ketone,acmc-1b06x
292625
pentadecan-7-one
CCCCCCCCC(=O)CCCCCC
Quantity 25g
Melting Point 30°C to 32°C
Formula Weight 226.4
Percent Purity 98%
Chemical Name or Material 7-Pentadecanone

TSCA : No

Recommended Storage : Ambient temperatures