Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions Available on GSA/VA Contract for Federal Government customers only.

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions.

Manufacturer: Thermo Scientific™ 7590C

 View more versions of this product

Catalog No. 09-980-382

  Check Availability
Add to Cart

Disclaimers:

Not intended for use in instrument calibration or diagnostic reagents.


    Specifications

    Diameter (Metric) 90μm
    Type Specialty Particles
    For Use With (Application) Fluid Mechanics
    Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
    Description Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems.
    Diameter (Metric) Particle 90µm
    Includes General product handling insert sheet
    Packaging 1000mL
    Particle Density 1.05g/cm3
    Quantity 1000mL
    Refractive Index 1.59 at 589nm (25°C)
    Stability Product stable for a minimum of 24 months
    Storage Requirements Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
    Concentration 10%
    Density 1.05g/cm3

    Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions, useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems.

    Suspensions of large copolymer microspheres are composed of polystyrene polymer, cross-linked with 4 to 8% divinylbenzene (DVB). The particles are chemically inert and can be washed with alcohol and vacuum or air dried. The polymer density is 1.05g/cm3 and the index of refraction is 1.59 at 589nm. They are packaged as aqueous suspensions at 10% solids by weight.

    Ordering Information:

    15mL, 100mL and 1000mL bottle sizes available. 15mL (“A” Part No. bottles), 100mL (“B” bottles) and 1000mL (“C” bottles) are packaged to order; example (Part No. 7503B and 7503C). Please allow 10 working days when ordering these fill volumes.

    Includes:

    Material Safety Data Sheet (MSDS) and general product handling insert sheet

    Recommended for:

    High concentration polymer particles > 3µm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications. Also useful as experimental particles for acoustical and optical analytical systems.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit