Learn More
Applied Biosystems™ Applied Biosystems™ 7500 Fast Dx Real-Time PCR Instrument, with tower computer
7500 Fast Dx RT-PCR Instrument with SDS Software is a real-time nucleic acid amplification and five-color fluorescence detection system available for in vitro diagnostic use.
Manufacturer: Applied Biosystems™ 4406985

Includes: Desktop Computer
Description
The 7500 Fast Dx Real-Time PCR Instrument delivers the performance required for high-quality results in a 96-well format.
- Accommodates tube strips, which can be capped immediately after pipetting each sample
- Fast mode completes runs in less than forty minutes
- Optional standard mode facilitates use of standard length real-time PCR assays without changing thermal cycling parameters
- 5-color variable excitation enables multiplex assays
- The 7500 Fast Dx Real-Time PCR Instrument is supported by our AB Assurance Service Plan for Diagnostics, which includes:
- Instrument Installation Qualification, Operation Qualification, and Performance Qualification (IQ⁄OQ⁄PQ)
- Spectral calibration and software setup
- Emergency repair, including parts and labor, and OQ⁄PQ after any major part change
- Planned maintenance every 12 months
Specifications
TAMRA, Cy3, FAM, SYBR Green, Texas Red, JOE, ROX, Cy5, NED, VIC | |
The package includes one real-time PCR instrument, one Dell tower computer, Primer Express Software, and Sequence Detection Software. Recommended operating conditions: Temperature: 15 to 30 °C (59 to 86 °F). Maximum change of less than 15 degrees Celsius (27 °F) per 24 hrs. Humidity: 20 to 80% relative humidity, noncondensing. | |
5.5°C | |
9 logs of linear dynamic range | |
96-well plate, 8-well strips | |
CCD Camera, Halogen light source, Five excitation and five emission filters | |
5.5°C⁄sec | |
10-30 µl | |
Fast mode: ± 3.5°C⁄sec, Standard mode: ± 1.6°C⁄sec | |
±0.5°C | |
±1°C | |
4 to 5 96-well plates per day | |
34 kg (75 lb) |
12 x 8-tube strips, 1 x 96-well plate (Fast) | |
Desktop Computer | |
Fast, Standard | |
Computer tower provided | |
7500 Fast Dx System | |
ROX (Separate Tube), Other Passive Reference Dye, ROX (Pre-mixed) | |
Distinguish between 5,000–10,000 genome equivalents (two-fold copy number difference) with 99.7% confidence | |
36 min (Fast), <2 hrs (Standard) | |
Down to 1 copy of human RNAse P gene | |
4.0-99.9 °C | |
Peltier-Based System | |
34 cm (13.39 in) (W) x 49 cm (19.29 in) (H) x 45 cm (17.72 in) (D) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.