Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 7712 Series PVC Coated Gloves

Catalog Number p-7232457
Click to view available options
Grip Design:
Rough
Smooth
Series:
7712 Series
7712R Series
This item is not returnable. View return policy
This item is not returnable. View return policy

PVC coated, multi-purpose gloves with soft cotton liner are designed for all industrial applications and protect against a range of hazards including chemicals, oils and abrasion.

Both styles are ideal for working with chemicals, oil, or grease in refining, mining, automotive subassembly, or construction

  • Fully coated PVC with smooth or rough finish
  • Cotton jersey lined Sanitized™
  • Recyclable
  • Washable

Certifications/Compliance CFIA
Color Black
Latex Free Yes
Material PVC
Size Large
For Use With (Application) Chemical spray and treatment, coating preparation, laboratory, pharma and analysis, washing and cleaning
Glove Cuff Style Gauntlet
Grip Design Smooth
Liner Material Cotton
Lining Lined
Model 7712-10
Series 7712 Series
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit