Learn More
Mopec 78 Inch Pedestal Style Autopsy Table
Includes all of the features necessary to efficiently perform an autopsy. Mopec™ 78 Inch Pedestal Style Autopsy Table is a pedestal designed table that includes standard downdraft ventilation system and is ideal for pediatric autopsies.
Supplier: Mopec CE400
Description
- Stainless steel pedestal autopsy table with down draft ventilation
- Tabletop is 14 gauge stainless steel with #4 satin finish
- Built-in large integral sink
- 2 GFCI receptacles positioned on each side of table
- Hydro aspirator designed to drain into sink plumbed with water control valve vacuum breaker and clear tubing
- Hot and cold single lever mixing valve with swing spout faucet complete with vacuum breaker
- Deck mounted chrome plated brass sink spray with high pressure hose
- 3 removable perforated grid plates
- Grid pattern is perforated with 9.5mm diameter holes on 50.8mm centers
Warranty and Services
1 Year Warranty
Specifications
Autopsy Table | |
2 GFCI Receptables Positioned on each side of Table | |
78 in. | |
93.98 cm | |
76.2 cm |
Grid Plates | |
37 in. | |
30 in. | |
198.12 cm |