Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 7X™-O-Matic Solution

7X™-O-Matic Solution was developed for safe and effective cleaning of laboratory instruments and glassware under pressure, heat and agitation (machine wash)

$75.60 - $256.00

Specifications

Concentration 7 x
Format Liquid
Storage Requirements 15 to 30°C
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
ICN7667494
View Documents
MP Biomedicals™
097667494
4 x1 gal.
Case of 4 for $256.00
Add to cart
 
ICN7667495
View Documents
MP Biomedicals™
097667495
5 gal.
Each for $209.00
Add to cart
 
ICN7667493
View Documents
MP Biomedicals™
097667493
1 gal.
Each for $75.60
Add to cart
 
Description

Description

  • Developed for safe and effective cleaning of laboratory instruments and glassware
  • Formulated to be used in a machine washer to withstand heat, pressure and agitation
  • Powerful wetting and penetrating agent, which contains a non-ionic surfactant and a hydrotrope to complex phosphates
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available, and it is stable for long-term storage
  • Effective and safe
  • Eliminates residue leaving no interfering fluorescence
  • No etch to glass or plastic labware at any concentration
  • Completely drains from glassware in seconds
Specifications

Specifications

7 x
Liquid
15 to 30°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit