Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ 7X™-PF-O-Matic Detergent
7X™-PF-O-Matic Detergent is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.
Supplier: MP Biomedicals™ 097667595
Description
- Environmentally safe and phosphate free
- Developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
- Powerful wetting and penetrating agent, designed to clean under pressure, heat and agitation
- Behaves and remains mostly suds-free when agitated
- Several convenient package sizes are available
- Stable for long-term storage
- Completely drains from glassware in seconds
- Eliminates residue leaving no interfering fluorescence
- pH neutral, and no need for pH adjustment at any concentration
- No etch to glass or plastic labware at any concentration
Specifications
Liquid | |
Dark Yellow | |
15°C to 30°C | |
5 gal. |
7 to 8 | |
7X | |
Use on Glass, Plastic, automatic washers |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction