Learn More
Seedburo Equipment CompanySupplier Diversity Partner 8/64"X 3/4" OBLONG STEEL SIEVE DFS Item
Manufacturer:  Seedburo Equipment CompanySupplier Diversity Partner 0100
8/64"X 3/4" OBLONG STEEL SIEVESeedburo Dockage Sieves are manufactured to comply with the specification of the U.S. Department of Agriculture. They are regularly used by federal grain supervision offices, state grain inspection departments and licensed grain inspectors. Most sieves are available in commercial or precision grades. All federal grain inspectors use precision sieves, but for most applications the commercial sieves will suffice.
Catalog No. NC1468435
Please call customer service for current pricing. 1-800-766-7000