Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Amino-1-naphthalenesulfonic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
82-75-7
C10H9NO3S
223.246
MFCD00035730
CYJJLCDCWVZEDZ-UHFFFAOYSA-N
peri acid,8-amino-1-naphthalenesulfonic acid,1-naphthylamine-8-sulfonic acid,1-naphthalenesulfonic acid, 8-amino,8-aminonaphthalene-1-sulphonic acid,naphthylaminemonosulfonic acid s,1-amino-8-naphthalene sulfonate,1-aminonaphthalene-8-sulfonic acid,schollkopf's acid van,schollkopf's acid
6722
8-aminonaphthalene-1-sulfonic acid
C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O
82-75-7
C10H9NO3S
223.246
MFCD00035730
CYJJLCDCWVZEDZ-UHFFFAOYSA-N
peri acid,8-amino-1-naphthalenesulfonic acid,1-naphthylamine-8-sulfonic acid,1-naphthalenesulfonic acid, 8-amino,8-aminonaphthalene-1-sulphonic acid,naphthylaminemonosulfonic acid s,1-amino-8-naphthalene sulfonate,1-aminonaphthalene-8-sulfonic acid,schollkopf's acid van,schollkopf's acid
6722
8-aminonaphthalene-1-sulfonic acid
C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O
Color Gray-Red
Quantity 500g
Formula Weight 223.25
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material 8-Amino-1-naphthalenesulfonic Acid

EINECSNumber : (4)-0491

TSCA : Yes