Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Amino-2-naphthalenesulfonic Acid 97.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
119-28-8
C10H9NO3S
223.246
MFCD00004032
QEZZCWMQXHXAFG-UHFFFAOYSA-N
8-amino-2-naphthalenesulfonic acid,1,7-cleve's acid,1-naphthylamine-7-sulfonic acid,2-naphthalenesulfonic acid, 8-amino,cleve's theta-acid,1-amino-7-sulfonaphthalene,1-aminonaphthalene-7-sulfonic acid,8-naphthylamine-2-sulfonic acid,1-amino-7-naphthalenesulfonic acid,1-amino-naphthalene-7-sulphonic acid
8386
8-aminonaphthalene-2-sulfonic acid
C1=CC2=C(C=C(C=C2)S(=O)(=O)O)C(=C1)N
119-28-8
C10H9NO3S
223.246
MFCD00004032
QEZZCWMQXHXAFG-UHFFFAOYSA-N
8-amino-2-naphthalenesulfonic acid,1,7-cleve's acid,1-naphthylamine-7-sulfonic acid,2-naphthalenesulfonic acid, 8-amino,cleve's theta-acid,1-amino-7-sulfonaphthalene,1-aminonaphthalene-7-sulfonic acid,8-naphthylamine-2-sulfonic acid,1-amino-7-naphthalenesulfonic acid,1-amino-naphthalene-7-sulphonic acid
8386
8-aminonaphthalene-2-sulfonic acid
C1=CC2=C(C=C(C=C2)S(=O)(=O)O)C(=C1)N
Color Undesignated
Quantity 500g
Formula Weight 223.25
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (HPLC,T)
Chemical Name or Material 8-Amino-2-naphthalenesulfonic Acid

EINECSNumber : (4)-0491

TSCA : Yes