Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Amino-2-naphthol, 94%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
118-46-7
C10H9NO
159.18
MFCD00004031
KVHHMYZBFBSVDI-UHFFFAOYSA-N
1-amino-7-naphthol,8-amino-2-naphthol,2-naphthalenol, 8-amino,1-amino-7-hydroxynaphthalene,7-hydroxy-1-naphthylamine,2-naphthol, 8-amino,8-amino-2-naphtol,8-aminonaphth-2-ol,1-amino-7-naphthol;,8-amino-2-naphthaleno
8358
8-aminonaphthalen-2-ol
C1=CC2=C(C=C(C=C2)O)C(=C1)N
118-46-7
C10H9NO
159.18
MFCD00004031
KVHHMYZBFBSVDI-UHFFFAOYSA-N
1-amino-7-naphthol,8-amino-2-naphthol,2-naphthalenol, 8-amino,1-amino-7-hydroxynaphthalene,7-hydroxy-1-naphthylamine,2-naphthol, 8-amino,8-amino-2-naphtol,8-aminonaphth-2-ol,1-amino-7-naphthol;,8-amino-2-naphthaleno
8358
8-aminonaphthalen-2-ol
C1=CC2=C(C=C(C=C2)O)C(=C1)N
Quantity 25g
Infrared Spectrum Authentic
Flash Point >364°C
Melting Point 204.0°C to 207.0°C
Color Brown to Gray
Packaging Glass bottle
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 92% min. (HPLC)
Beilstein 13, 685
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 159.18
Physical Form Powder or Crystals
Percent Purity 94%
Chemical Name or Material 8-Amino-2-naphthol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-252-4