Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Aminoquinoline, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
578-66-5
C9H8N2
144.18
MFCD00006809
WREVVZMUNPAPOV-UHFFFAOYSA-N
8-aminoquinoline,8-quinolinamine,8-quinolylamine,quinoline, 8-amino,quinolin-8-ylamine,unii-u34eav21tg,8-amino quinoline,ccris 1683,u34eav21tg,8-amino-quinoline
11359
quinolin-8-amine
C1=CC2=C(C(=C1)N)N=CC=C2
578-66-5
C9H8N2
144.18
MFCD00006809
WREVVZMUNPAPOV-UHFFFAOYSA-N
8-aminoquinoline,8-quinolinamine,8-quinolylamine,quinoline, 8-amino,quinolin-8-ylamine,unii-u34eav21tg,8-amino quinoline,ccris 1683,u34eav21tg,8-amino-quinoline
11359
quinolin-8-amine
C1=CC2=C(C(=C1)N)N=CC=C2
Infrared Spectrum Authentic
Melting Point 62.0°C to 65.0°C
Quantity 50g
Packaging Glass bottle
Boiling Point 174.0°C (26.0 mmHg)
Color Undesignated
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Beilstein 22, 450
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, insoluble in ether
Formula Weight 144.18
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material 8-Aminoquinoline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause damage to organs.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning