Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Aminoquinoline N-Oxide 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
92339-84-9
C9H8N2O
160.176
VAPLDXAYDSGTPC-UHFFFAOYSA-N
594833
1-oxidoquinolin-1-ium-8-amine
C1=CC2=C(C(=C1)N)[N+](=CC=C2)[O-]
92339-84-9
C9H8N2O
160.176
VAPLDXAYDSGTPC-UHFFFAOYSA-N
594833
1-oxidoquinolin-1-ium-8-amine
C1=CC2=C(C(=C1)N)[N+](=CC=C2)[O-]
Quantity 5g
Color Yellow
Melting Point 110°C
Formula Weight 160.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 8-Aminoquinoline N-Oxide