Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Aminoquinoline 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
100g
578-66-5
C9H8N2
144.177
MFCD00006809
WREVVZMUNPAPOV-UHFFFAOYSA-N
8-aminoquinoline,8-quinolinamine,8-quinolylamine,quinoline, 8-amino,quinolin-8-ylamine,unii-u34eav21tg,8-amino quinoline,ccris 1683,u34eav21tg,8-amino-quinoline
11359
quinolin-8-amine
C1=CC2=C(C(=C1)N)N=CC=C2
578-66-5
C9H8N2
144.177
MFCD00006809
WREVVZMUNPAPOV-UHFFFAOYSA-N
8-aminoquinoline,8-quinolinamine,8-quinolylamine,quinoline, 8-amino,quinolin-8-ylamine,unii-u34eav21tg,8-amino quinoline,ccris 1683,u34eav21tg,8-amino-quinoline
11359
quinolin-8-amine
C1=CC2=C(C(=C1)N)N=CC=C2
Boiling Point 145°C
Color Undesignated
Quantity 25g
Melting Point 65°C
Formula Weight 144.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 8-Aminoquinoline

RTECSNumber : VA9627000

TSCA : Yes