Alfa Aesar™ 8-Bromo-1,6-naphthyridin-2(1H)-one, 98%

Manufacturer: Alfa Aesar™ H5004503

Quantity 1g

 View more versions of this product

Catalog No. AAH5004503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 902837-41-6
    Molecular Formula C8H5BrN2O
    Formula Weight 225.05
    MDL Number MFCD08060938
    Synonym 8-Bromo-2-hydroxy-1,6-naphthyridine; 6-Bromo-1,6-naphthyridin-2-ol
    Chemical Name or Material 8-Bromo-1,6-naphthyridin-2(1H)-one

    Specifications

    Formula Weight 225.05
    Molecular Formula C8H5BrN2O
    MDL Number MFCD08060938
    Synonym 8-Bromo-2-hydroxy-1,6-naphthyridine; 6-Bromo-1,6-naphthyridin-2-ol
    Quantity 1g
    CAS 902837-41-6
    Chemical Name or Material 8-Bromo-1,6-naphthyridin-2(1H)-one
    Assay 98%

    Description

    Applications
    8-Bromo-1,6-naphthyridin-2(1H)-one is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit