Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromo-1-octanol, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
50816-19-8
C8H17BrO
209.13
GMXIEASXPUEOTG-UHFFFAOYSA-N
8-bromo-1-octanol,1-octanol, 8-bromo,8-bromooctanol,octamethylene bromohydrin,8-bromo octanol,1-bromooctan-8-ol,8-bromo-1-actanol,8-bromooctane-1-ol,1-bromo-octan-8-ol,8-bromo-octan-1-ol
162607
8-bromooctan-1-ol
C(CCCCBr)CCCO
50816-19-8
C8H17BrO
209.13
GMXIEASXPUEOTG-UHFFFAOYSA-N
8-bromo-1-octanol,1-octanol, 8-bromo,8-bromooctanol,octamethylene bromohydrin,8-bromo octanol,1-bromooctan-8-ol,8-bromo-1-actanol,8-bromooctane-1-ol,1-bromo-octan-8-ol,8-bromo-octan-1-ol
162607
8-bromooctan-1-ol
C(CCCCBr)CCCO
Packaging Glass Bottle
Boiling Point 79.0°C to 80.0°C (0.1mmHg)
Density 1.2200g/mL
Flash Point >110°C
Quantity 1g
Specific Gravity 1.22
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Formula Weight 209.13
Percent Purity 95%
Chemical Name or Material 8-Bromo-1-octanol
Show More Show Less

EINECSNumber : 256-785-7