Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 8-Bromo-1-octene, 97%

Click to view available options
Quantity:
1g
5g
2695-48-9
C8H15Br
191.112
MFCD00000275
SNMOMUYLFLGQQS-UHFFFAOYSA-N
8-bromo-1-octene,1-octene, 8-bromo,7-octenyl bromide,8-bromooctene,8-azaquanine,bromo-8 octene-1,8-bromo-oct-1-ene,acmc-1cmcf,oct-7-en-1-yl bromide,8-bromo-1-octene
75907
8-bromooct-1-ene
C=CCCCCCCBr
2695-48-9
C8H15Br
191.112
MFCD00000275
SNMOMUYLFLGQQS-UHFFFAOYSA-N
8-bromo-1-octene,1-octene, 8-bromo,7-octenyl bromide,8-bromooctene,8-azaquanine,bromo-8 octene-1,8-bromo-oct-1-ene,acmc-1cmcf,oct-7-en-1-yl bromide,8-bromo-1-octene
75907
8-bromooct-1-ene
C=CCCCCCCBr
Applications
8-Bromo-1-octene has been used in preparation of polymerizable ligand, required for the synthesis of quantum dot-labeled polymer beads. Grignard reagent derived from 8-bromo-1-octene has been used in the synthesis of (2S,3S,5R)-5-[(1R)-1-hydroxy-9-decenyl]-2-pentyltetrahydro-3-furanol. It has been used as building block in natural product synthesis. Also used for the synthesis of more complex pharmaceutical compounds, such as Dithienogermole (DTG) derivatives with varying alkyl chain lengths and pendant functionalities.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Refractive Index 1.4670
Density 1.139g/mL
Color Undesignated
Quantity 1g
Boiling Point 92°C (24mmHg)
Linear Formula Br(CH2)6CH=CH2
Beilstein 2234481
Formula Weight 191.11
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 8-Bromo-1-octene
Show More Show Less

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning