Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 8-Bromo-2-chloro-6-fluoroquinazoline, 95%

Click to view available options
Quantity:
250mg
1g
953039-63-9
C8H3BrClFN2
261.478
MFCD18382554
XTEWVYPXKPYPRE-UHFFFAOYSA-N
8-bromo-2-chloro-6-fluoro-quinazoline,quinazoline, 8-bromo-2-chloro-6-fluoro
59199299
8-bromo-2-chloro-6-fluoroquinazoline
C1=C(C=C2C=NC(=NC2=C1Br)Cl)F
953039-63-9
C8H3BrClFN2
261.478
MFCD18382554
XTEWVYPXKPYPRE-UHFFFAOYSA-N
8-bromo-2-chloro-6-fluoro-quinazoline,quinazoline, 8-bromo-2-chloro-6-fluoro
59199299
8-bromo-2-chloro-6-fluoroquinazoline
C1=C(C=C2C=NC(=NC2=C1Br)Cl)F
Quantity 250mg
Beilstein 15583920
Formula Weight 261.48
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 8-Bromo-2-chloro-6-fluoroquinazoline

storageNote1 : -30° to -10°C