Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromoquinoline, 96%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass Bottle in Plastic Container
16567-18-3
C9H6BrN
208.058
MFCD00191859
PIWNKSHCLTZKSZ-UHFFFAOYSA-N
quinoline, 8-bromo,8-bromo-quinoline,8-bromoquinoline,pubchem5816,8-bromquinoline,acmc-209dt3,ksc181q5t,buttpark 33\04-57
140109
8-bromoquinoline
C1=CC2=C(C(=C1)Br)N=CC=C2
16567-18-3
C9H6BrN
208.058
MFCD00191859
PIWNKSHCLTZKSZ-UHFFFAOYSA-N
quinoline, 8-bromo,8-bromo-quinoline,8-bromoquinoline,pubchem5816,8-bromquinoline,acmc-209dt3,ksc181q5t,buttpark 33\04-57
140109
8-bromoquinoline
C1=CC2=C(C(=C1)Br)N=CC=C2
Infrared Spectrum Authentic
Quantity 500mg
Color Brown-Yellow to Yellow
Refractive Index 1.6675 to 1.6700 (20°C, 589nm)
Packaging Glass Bottle in Plastic Container
Density 1.5940g/mL
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Beilstein 20, 365
Formula Weight 208.06
Physical Form Liquid After Melting
Percent Purity ≥95%
Chemical Name or Material 8-Bromoquinoline
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.