Learn More
National Safety Apparel™ 8 Cal ArcGuard™ Protera Arc Flash Kit with FR Coverall (10)
With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
Manufacturer:  National Safety Apparel™ KIT2CVPR084X10
 View more versions of this product
Catalog No. 17-001-1105
Add to cart
Description
Description
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. DuPont™ Protera™
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper
  • Made in the USA
Specifications
Specifications
Arc Rating: 8 cal/cm², CAT 2, NFPA 70 E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Open
DuPont™ Protera™
4X-Large
Zipper
Khaki
Stitched
None