Learn More
National Safety Apparel™ 8 Cal ArcGuard™ Protera Arc Flash Kit with FR Coverall (11)
With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
$785.50 - $845.51
Specifications
Seam Type Stitched
Material DuPont™ Protera™
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1076
View Documents
National Safety Apparel™
KIT2CVPR08SM11
Small
Each for $785.50
Add to cart
17-001-1081
View Documents
National Safety Apparel™
KIT2CVPR08MD11
Medium
Each for $785.50
Add to cart
17-001-1086
View Documents
National Safety Apparel™
KIT2CVPR08LG11
Large
Each for $785.50
Add to cart
17-001-1091
View Documents
National Safety Apparel™
KIT2CVPR08XL11
X-Large
Each for $785.50
Add to cart
17-001-1096
View Documents
National Safety Apparel™
KIT2CVPR082X11
2X-Large
Each for $805.50
Add to cart
17-001-1101
View Documents
National Safety Apparel™
KIT2CVPR083X11
3X-Large
Each for $805.50
Add to cart
17-001-1106
View Documents
National Safety Apparel™
KIT2CVPR084X11
4X-Large
Each for $825.51
Add to cart
17-001-1111
View Documents
National Safety Apparel™
KIT2CVPR085X11
5X-Large
Each for $845.51
Add to cart
Description
Description
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. DuPont™ Protera™
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper
  • Made in the USA
Specifications
Specifications
Stitched
DuPont™ Protera™