Learn More
National Safety Apparel™ 8 Cal ArcGuard™ Protera Arc Flash Kit with FR Coverall (12)
With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
$785.50 - $845.51
Specifications
Seam Type Stitched
Material DuPont™ Protera™
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1077
View Documents
National Safety Apparel™
KIT2CVPR08SM12
Small
Each for $785.50
Add to cart
17-001-1082
View Documents
National Safety Apparel™
KIT2CVPR08MD12
Medium
Each for $785.50
Add to cart
17-001-1087
View Documents
National Safety Apparel™
KIT2CVPR08LG12
Large
Each for $785.50
Add to cart
17-001-1092
View Documents
National Safety Apparel™
KIT2CVPR08XL12
X-Large
Each for $785.50
Add to cart
17-001-1097
View Documents
National Safety Apparel™
KIT2CVPR082X12
2X-Large
Each for $805.50
Add to cart
17-001-1102
View Documents
National Safety Apparel™
KIT2CVPR083X12
3X-Large
Each for $805.50
Add to cart
17-001-1107
View Documents
National Safety Apparel™
KIT2CVPR084X12
4X-Large
Each for $825.51
Add to cart
17-001-1112
View Documents
National Safety Apparel™
KIT2CVPR085X12
5X-Large
Each for $845.51
Add to cart
Description
Description
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. DuPont™ Protera™
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper
  • Made in the USA
Specifications
Specifications
Stitched
DuPont™ Protera™