Learn More
National Safety Apparel™ 8 Cal ArcGuard™ Protera Arc Flash Kit with FR Coverall and Balaclava (08)
Arc flash kit with Pureview and FR coverall in protera and balaclava
$821.76 - $881.77
Specifications
Seam Type Stitched
Material DuPont™ Protera™
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1113
View Documents
National Safety Apparel™
KIT2CVPR08BSM08
Small
Each for $821.76
Add to cart
17-001-1118
View Documents
National Safety Apparel™
KIT2CVPR08BMD08
Medium
Each for $821.76
Add to cart
17-001-1123
View Documents
National Safety Apparel™
KIT2CVPR08BLG08
Large
Each for $821.76
Add to cart
17-001-1128
View Documents
National Safety Apparel™
KIT2CVPR08BXL08
X-Large
Each for $821.76
Add to cart
17-001-1133
View Documents
National Safety Apparel™
KIT2CVPR08B2X08
2X-Large
Each for $841.76
Add to cart
17-001-1138
View Documents
National Safety Apparel™
KIT2CVPR08B3X08
3X-Large
Each for $841.76
Add to cart
17-001-1143
View Documents
National Safety Apparel™
KIT2CVPR08B4X08
4X-Large
Each for $861.77
Add to cart
17-001-1148
View Documents
National Safety Apparel™
KIT2CVPR08B5X08
5X-Large
Each for $881.77
Add to cart
Description
Description
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. DuPont™ Protera™
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper
  • Made in the USA
Specifications
Specifications
Stitched
DuPont™ Protera™