Learn More
National Safety Apparel™ 8 Cal ArcGuard™ Protera Arc Flash Kit with FR Coverall and Balaclava (11)
Arc flash kit with Pureview and FR coverall in protera and balaclava
$821.76 - $881.77
Specifications
Seam Type Stitched
Material DuPont™ Protera™
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1116
View Documents
National Safety Apparel™
KIT2CVPR08BSM11
Small
Each for $821.76
Add to cart
17-001-1121
View Documents
National Safety Apparel™
KIT2CVPR08BMD11
Medium
Each for $821.76
Add to cart
17-001-1126
View Documents
National Safety Apparel™
KIT2CVPR08BLG11
Large
Each for $821.76
Add to cart
17-001-1131
View Documents
National Safety Apparel™
KIT2CVPR08BXL11
X-Large
Each for $821.76
Add to cart
17-001-1136
View Documents
National Safety Apparel™
KIT2CVPR08B2X11
2X-Large
Each for $841.76
Add to cart
17-001-1141
View Documents
National Safety Apparel™
KIT2CVPR08B3X11
3X-Large
Each for $841.76
Add to cart
17-001-1146
View Documents
National Safety Apparel™
KIT2CVPR08B4X11
4X-Large
Each for $861.77
Add to cart
17-001-1151
View Documents
National Safety Apparel™
KIT2CVPR08B5X11
5X-Large
Each for $881.77
Add to cart
Description
Description
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. DuPont™ Protera™
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper
  • Made in the USA
Specifications
Specifications
Stitched
DuPont™ Protera™