Learn More
National Safety Apparel™ 8 Cal Protera FR Coveralls 30 in. Inseam
Coveralls with two-way covered zipper down front for easy entry
$251.15 - $334.85
Specifications
Material Modacrylic, Aramid fiber, Anti-Static
Seam Type Stitched
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-381
View Documents
National Safety Apparel™
C88LISM30
Small
Each for $251.15
Add to cart
17-001-382
View Documents
National Safety Apparel™
C88LIMD30
Medium
Each for $251.15
Add to cart
17-001-383
View Documents
National Safety Apparel™
C88LILG30
Large
Each for $251.15
Add to cart
17-001-384
View Documents
National Safety Apparel™
C88LIXL30
X-Large
Each for $251.15
Add to cart
17-001-385
View Documents
National Safety Apparel™
C88LI2X30
2X-Large
Each for $276.58
Add to cart
17-001-386
View Documents
National Safety Apparel™
C88LI3X30
3X-Large
Each for $276.58
Add to cart
17-001-387
View Documents
National Safety Apparel™
C88LI4X30
4X-Large
Each for $304.15
Add to cart
17-001-388
View Documents
National Safety Apparel™
C88LI5X30
5X-Large
Each for $334.85
Add to cart
Description
Description
  • Arc Rating: 8.5 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. Protera™
  • Dual Hazard
  • Two-way covered zipper down front for easy entry
  • Comfortable, sewn-in elastic waist
  • Chest pockets with flaps and hidden snap closures
  • Reinforced front and back pockets
  • Convenient side pass-through pockets for access to pant pockets
  • Leg openings made to fit over work boots
  • Action back allows for full range of motion
  • All metal covered to ensure no contact with skin
  • Made in the USA
Specifications
Specifications
Modacrylic, Aramid fiber, Anti-Static
Stitched