Learn More
National Safety Apparel™ 8 Cal Protera FR Coveralls 32 in. Inseam
Coveralls with two-way covered zipper down front for easy entry
$251.15 - $334.85
Specifications
Material Modacrylic, Aramid fiber, Anti-Static
Seam Type Stitched
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-389
View Documents
National Safety Apparel™
C88LISM32
Small
Each for $251.15
Add to cart
17-001-390
View Documents
National Safety Apparel™
C88LIMD32
Medium
Each for $251.15
Add to cart
17-001-391
View Documents
National Safety Apparel™
C88LILG32
Large
Each for $251.15
Add to cart
17-001-392
View Documents
National Safety Apparel™
C88LIXL32
X-Large
Each for $251.15
Add to cart
17-001-393
View Documents
National Safety Apparel™
C88LI2X32
2X-Large
Each for $276.58
Add to cart
17-001-394
View Documents
National Safety Apparel™
C88LI3X32
3X-Large
Each for $276.58
Add to cart
17-001-395
View Documents
National Safety Apparel™
C88LI4X32
4X-Large
Each for $304.15
Add to cart
17-001-396
View Documents
National Safety Apparel™
C88LI5X32
5X-Large
Each for $334.85
Add to cart
Description
Description
  • Arc Rating: 8.5 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. Protera™
  • Dual Hazard
  • Two-way covered zipper down front for easy entry
  • Comfortable, sewn-in elastic waist
  • Chest pockets with flaps and hidden snap closures
  • Reinforced front and back pockets
  • Convenient side pass-through pockets for access to pant pockets
  • Leg openings made to fit over work boots
  • Action back allows for full range of motion
  • All metal covered to ensure no contact with skin
  • Made in the USA
Specifications
Specifications
Modacrylic, Aramid fiber, Anti-Static
Stitched