Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ 8 Cal UltraSoft Khaki Coverall 30 in. Inseam

For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability.  With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability

Manufacturer:  National Safety Apparel™ C88UK5X30

 View more versions of this product

Catalog No. 17-001-516


Add to cart

Description

Description

  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 7 oz. Westex™ UltraSoft™
  • Action back allows for full range of motion
  • All metal covered to ensure no contact with skin
  • Chest pockets with flaps and hidden snap closures
  • Comfortable sewn-in elastic waist for better fit
  • Convenient side pass-through pockets for access to pant pockets
  • Covered zipper front for easy entry
  • FR fabric will not melt, drip or ignite
  • FR protection from electrical arc flash and flash fire hazards
  • Home laundry
  • Leg openings made to fit over work boots
  • Two front pockets with pass-through for access to pants pockets
  • Made in the USA
Specifications

Specifications

Arc Rating: 8 cal/cm², CAT 2, NFPA 70 E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Open
Cotton, Nylon
5X-Large
Zipper
Khaki
Stitched
None
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit