Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ 8-Channel Micropipette
The 8-Channel Micropipette is fully autoclavable and recommended for a wide variety of applications such as ELISA, molecular screening, kinetic studies, and DNA amplifications.
Supplier: United Scientific™ PMC8100
Description
- Multichannel micropipette.
- Two-stop plunger.
- Built-in tip ejector.
- Autoclavable tip cone.
- Click-stop adjustable digital display.
- Easy to calibrate, maintain, and disassemble for autoclaving.
- Manufactured in an ISO 9001:2008 certified facility.
- Each unit calibrated according to ISO 8655 standards and includes a calibration certificate.
Specifications
±3.0% at 10.0 μL, ±1.0% at 50.0 μL, ±1.0% at 100.0 μL | |
No | |
White/Blue | |
Micro Tip 200 μL | |
Plastic | |
8 | |
Non-sterile | |
10 to 100 μL | |
Micropipette Tips | |
≤1.5% at 10.0 μL, ≤0.5% at 50.0 μL, ≤0.5% at 100.0 μL |
200 μL | |
Autoclavable | |
Reusable | |
Ergonomic | |
PMC8-100 | |
No | |
Micropipette | |
Electrophoresis, PCR | |
Eight tips, One pipette holder | |
1 Year |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction