Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-chloro-1-octanol, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
23144-52-7
C8H17ClO
164.67
YDFAJMDFCCJZSI-UHFFFAOYSA-N
8-chloro-1-octanol,1-octanol, 8-chloro,8-chlorooctanol,8-chloro-octan-1-ol,8-chloro-1-n-octanol,8-chlor-1-octanol,8-chlor-octan-1-ol,acmc-1cr4x
90012
8-chlorooctan-1-ol
C(CCCCCl)CCCO
23144-52-7
C8H17ClO
164.67
YDFAJMDFCCJZSI-UHFFFAOYSA-N
8-chloro-1-octanol,1-octanol, 8-chloro,8-chlorooctanol,8-chloro-octan-1-ol,8-chloro-1-n-octanol,8-chlor-1-octanol,8-chlor-octan-1-ol,acmc-1cr4x
90012
8-chlorooctan-1-ol
C(CCCCCl)CCCO
Density 0.9760g/mL
Refractive Index 1.4575 to 1.4595
Flash Point >113°C
Packaging Glass Bottle
Boiling Point 129°C to 130°C (15.0mmHg)
Specific Gravity 0.976
Color Colorless
Infrared Spectrum Authentic
Quantity 5mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula Cl(CH2)8OH
Formula Weight 164.67
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 8-Chloro-1-octanol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 245-451-6