Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline, 97%

Click to view available options
Quantity:
1g
5g
18706-22-4
C10H5ClF3NO
247.601
MFCD00276589
SBVZSZNYPRWWNN-UHFFFAOYSA-N
8-chloro-4-hydroxy-2-trifluoromethyl quinoline,8-chloro-2-trifluoromethyl quinolin-4-ol,8-chloro-2-trifluoromethyl-1h-quinolin-4-one,8-chloro-2-trifluoromethyl-4-quinolinol,4 1h-quinolinone, 8-chloro-2-trifluoromethyl,maybridge1_004966,pubchem5940,acmc-1bvi2,8-chloro-2-trifluoromethyl-quinolin-4-ol,8-chloro-4-hydroxy-2-trifluoromethylquinoline
2736611
8-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C(=C1)Cl)NC(=CC2=O)C(F)(F)F
18706-22-4
C10H5ClF3NO
247.601
MFCD00276589
SBVZSZNYPRWWNN-UHFFFAOYSA-N
8-chloro-4-hydroxy-2-trifluoromethyl quinoline,8-chloro-2-trifluoromethyl quinolin-4-ol,8-chloro-2-trifluoromethyl-1h-quinolin-4-one,8-chloro-2-trifluoromethyl-4-quinolinol,4 1h-quinolinone, 8-chloro-2-trifluoromethyl,maybridge1_004966,pubchem5940,acmc-1bvi2,8-chloro-2-trifluoromethyl-quinolin-4-ol,8-chloro-4-hydroxy-2-trifluoromethylquinoline
2736611
8-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C(=C1)Cl)NC(=CC2=O)C(F)(F)F
Quantity 1g
Melting Point 138°C to 140°C
Formula Weight 247.61
Percent Purity 97%
Chemical Name or Material 8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning