Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 8-Chlorotheophylline, 99%

Manufacturer:  Alfa Aesar™ A1240818

Quantity 50g

 View more versions of this product

Catalog No. AAA1240818

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
8-Chlorotheophylline is a xanthine alkaloid derivative. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in sodium hydroxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

85-18-7
214.61
203068
8-Chlorotheophylline
C7H7ClN4O2
MFCD00005581
8-Chloro-1,3-dimethyl-2,6-purinedione

Specifications

≈290°C (dec.)
85-18-7
214.61
203068
8-Chlorotheophylline
50g
C7H7ClN4O2
MFCD00005581
8-Chloro-1,3-dimethyl-2,6-purinedione
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XH5063000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit