Alfa Aesar™ 8-Chlorotheophylline, 99%

Manufacturer: Alfa Aesar™ A124080B

Quantity 1000g

 View more versions of this product

Catalog No. AAA124080B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85-18-7
    Molecular Formula C7H7ClN4O2
    Formula Weight 214.61
    MDL Number MFCD00005581
    Beilstein 203068
    Synonym 8-Chloro-1,3-dimethyl-2,6-purinedione
    Chemical Name or Material 8-Chlorotheophylline

    Specifications

    Formula Weight 214.61
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00005581
    CAS 85-18-7
    Melting Point ≈290°C (dec.)
    Molecular Formula C7H7ClN4O2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    RTECS Number XH5063000
    Quantity 1000g
    Beilstein 203068
    Synonym 8-Chloro-1,3-dimethyl-2,6-purinedione
    Chemical Name or Material 8-Chlorotheophylline
    Assay 99%

    Description

    Applications
    8-Chlorotheophylline is a xanthine alkaloid derivative. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

    Solubility
    Soluble in sodium hydroxide.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit