Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 8-Ethoxyquinoline-5-sulfonic acid sodium salt hydrate, 97%

Click to view available options
Quantity:
10g
50g
250g
80789-76-0
C11H12NNaO5S
293.269
MFCD00149478
BSVDXMZRSQZXAH-UHFFFAOYSA-M
sodium 8-ethoxyquinoline-5-sulfonate hydrate,8-ethoxyquinoline-5-sulfonic acid sodium salt hemihydroate,actinoquinol sodium salt,8-ethoxyquinoline-5-sulfonic acid sodium salt hydrate,8-ethoxy-5-quinolinesulfonic acid sodium salt monohydrate,acmc-2097kw,sodium 8-ethoxyquinoline-5-sulfonate monohydrate,8-ethoxyquinoline-5-sulfonic acid sodium salt hemihydrate
51063116
sodium;8-ethoxyquinoline-5-sulfonate;hydrate
CCOC1=C2C(=C(C=C1)S(=O)(=O)[O-])C=CC=N2.O.[Na+]
80789-76-0
C11H12NNaO5S
293.269
MFCD00149478
BSVDXMZRSQZXAH-UHFFFAOYSA-M
sodium 8-ethoxyquinoline-5-sulfonate hydrate,8-ethoxyquinoline-5-sulfonic acid sodium salt hemihydroate,actinoquinol sodium salt,8-ethoxyquinoline-5-sulfonic acid sodium salt hydrate,8-ethoxy-5-quinolinesulfonic acid sodium salt monohydrate,acmc-2097kw,sodium 8-ethoxyquinoline-5-sulfonate monohydrate,8-ethoxyquinoline-5-sulfonic acid sodium salt hemihydrate
51063116
sodium;8-ethoxyquinoline-5-sulfonate;hydrate
CCOC1=C2C(=C(C=C1)S(=O)(=O)[O-])C=CC=N2.O.[Na+]
Sensitivity Hygroscopic
Melting Point >280°C
Quantity 250g
Formula Weight 275.26 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 8-Ethoxyquinoline-5-sulfonic acid sodium salt hydrate

EINECSNumber : 230-651-8

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen