Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Fluoroquinolin-4-ol, 97%

Click to view available options
Quantity:
1g
63010-71-9
C9H6FNO
163.151
MFCD00272395
HTELWJVKZKSAQI-UHFFFAOYSA-N
8-fluoroquinolin-4-ol,8-fluoro-4-hydroxyquinoline,8-fluoroquinolin-4 1h-one,4-hydroxy-8-fluoroquinoline,4-quinolinol, 8-fluoro,8-fluoro-4-quinolinol,8-fluoro-quinolin-4-ol,pubchem5899,acmc-1b6pg,8-fluoro-4-hydroxy quinoline
2737334
8-fluoro-1H-quinolin-4-one
C1=CC2=C(C(=C1)F)NC=CC2=O
63010-71-9
C9H6FNO
163.151
MFCD00272395
HTELWJVKZKSAQI-UHFFFAOYSA-N
8-fluoroquinolin-4-ol,8-fluoro-4-hydroxyquinoline,8-fluoroquinolin-4 1h-one,4-hydroxy-8-fluoroquinoline,4-quinolinol, 8-fluoro,8-fluoro-4-quinolinol,8-fluoro-quinolin-4-ol,pubchem5899,acmc-1b6pg,8-fluoro-4-hydroxy quinoline
2737334
8-fluoro-1H-quinolin-4-one
C1=CC2=C(C(=C1)F)NC=CC2=O
Melting Point 280°C to 282°C
Quantity 1g
Formula Weight 163.15
Percent Purity 97%
Chemical Name or Material 8-Fluoroquinolin-4-ol

GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement
P280-P301+P310-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger