Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 8-(Fmoc-amino)octanoic acid, 95%

Click to view available options
Quantity:
1g
5g
126631-93-4
C23H27NO4
381.472
MFCD00235890
QZQXRZXYWVQWAY-UHFFFAOYSA-N
n-fmoc-8-aminooctanoic acid,fmoc-8-aoc-oh,monascamine,monascorubramin,fmoc-8-aminocaprylic acid,unii-wq3ps53e2u,wq3ps53e2u,8-9h-fluoren-9-ylmethoxycarbonylamino octanoic acid,8-fmoc-amino-octanoic acid,8-9h-fluoren-9-yl methoxy carbonyl amino octanoic acid
2756091
8-(9H-fluoren-9-ylmethoxycarbonylamino)octanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCCC(=O)O
126631-93-4
C23H27NO4
381.472
MFCD00235890
QZQXRZXYWVQWAY-UHFFFAOYSA-N
n-fmoc-8-aminooctanoic acid,fmoc-8-aoc-oh,monascamine,monascorubramin,fmoc-8-aminocaprylic acid,unii-wq3ps53e2u,wq3ps53e2u,8-9h-fluoren-9-ylmethoxycarbonylamino octanoic acid,8-fmoc-amino-octanoic acid,8-9h-fluoren-9-yl methoxy carbonyl amino octanoic acid
2756091
8-(9H-fluoren-9-ylmethoxycarbonylamino)octanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCCC(=O)O
Quantity 5g
Color White
Beilstein 8286497
Formula Weight 381.47
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 8-(Fmoc-amino)octanoic acid

storageNote1 : 2° to 8°C