Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 8-Hydroxy-5-nitroquinoline, 96%

Click to view available options
Quantity:
5g
25g
100g
4008-48-4
C9H6N2O3
190.158
MFCD00006791
RJIWZDNTCBHXAL-UHFFFAOYSA-N
nitroxoline,5-nitroquinolin-8-ol,8-hydroxy-5-nitroquinoline,5-nitro-8-hydroxyquinoline,5-nitro-8-quinolinol,nitroxolin,noxibiol,5-nitrox,5-nitroxine,5-nitroks
19910
CHEBI:67121
C1=CC2=C(C=CC(=C2N=C1)O)[N+](=O)[O-]
4008-48-4
C9H6N2O3
190.158
MFCD00006791
RJIWZDNTCBHXAL-UHFFFAOYSA-N
nitroxoline,5-nitroquinolin-8-ol,8-hydroxy-5-nitroquinoline,5-nitro-8-hydroxyquinoline,5-nitro-8-quinolinol,nitroxolin,noxibiol,5-nitrox,5-nitroxine,5-nitroks
19910
CHEBI:67121
C1=CC2=C(C=CC(=C2N=C1)O)[N+](=O)[O-]
Melting Point 177°C to 182°C
Odor Odorless
Quantity 100g
Merck Index 14,6655
Formula Weight 190.16
Percent Purity 96%
Chemical Name or Material 8-Hydroxy-5-nitroquinoline

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 223-662-4

RTECSNumber : VC8225000

TSCA : No

Recommended Storage : Ambient temperatures