Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinaldine, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
826-81-3
C10H9NO
159.19
MFCD00006765
NBYLBWHHTUWMER-UHFFFAOYSA-N
8-hydroxyquinaldine,2-methyl-8-hydroxyquinoline,2-methyl-8-quinolinol,8-quinolinol, 2-methyl,2-methyloxine,8-hydroxy-2-methylquinoline,hydroxyquinaldine,8-hydroxyqinaldine,2-methyl-quinolin-8-ol,2-methyl 8-quinolonol
13224
2-methylquinolin-8-ol
CC1=NC2=C(C=CC=C2O)C=C1
826-81-3
C10H9NO
159.19
MFCD00006765
NBYLBWHHTUWMER-UHFFFAOYSA-N
8-hydroxyquinaldine,2-methyl-8-hydroxyquinoline,2-methyl-8-quinolinol,8-quinolinol, 2-methyl,2-methyloxine,8-hydroxy-2-methylquinoline,hydroxyquinaldine,8-hydroxyqinaldine,2-methyl-quinolin-8-ol,2-methyl 8-quinolonol
13224
2-methylquinolin-8-ol
CC1=NC2=C(C=CC=C2O)C=C1
Boiling Point 267°C
Flash Point 139°C
Melting Point 70°C to 74°C
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 21, 106
Solubility Information Solubility in water: insoluble
Formula Weight 159.19
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 8-Hydroxyquinaldine, 98%
Show More Show Less

EINECSNumber : 212-562-6

RTECSNumber : VC7920000

TSCA : TSCA