Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinoline-2-carboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1571-30-8
C10H7NO3
189.17
MFCD00168972
UHBIKXOBLZWFKM-UHFFFAOYSA-N
8-hydroxyquinaldic acid,quinaldic acid, 8-hydroxy,2-quinolinecarboxylic acid, 8-hydroxy,8-hydroxy-2-quinolinecarboxylic acid,8hc,acmc-20a4qz,5-22-05-00272 beilstein handbook reference,8-hydroxyquinolin-2-carboxylic acid,8-hydroxyquinoline-2-carbaxylicacid,quinolinecarboxylic acid, 8-hydroxy
74079
8-hydroxyquinoline-2-carboxylic acid
C1=CC2=C(C(=C1)O)N=C(C=C2)C(=O)O
1571-30-8
C10H7NO3
189.17
MFCD00168972
UHBIKXOBLZWFKM-UHFFFAOYSA-N
8-hydroxyquinaldic acid,quinaldic acid, 8-hydroxy,2-quinolinecarboxylic acid, 8-hydroxy,8-hydroxy-2-quinolinecarboxylic acid,8hc,acmc-20a4qz,5-22-05-00272 beilstein handbook reference,8-hydroxyquinolin-2-carboxylic acid,8-hydroxyquinoline-2-carbaxylicacid,quinolinecarboxylic acid, 8-hydroxy
74079
8-hydroxyquinoline-2-carboxylic acid
C1=CC2=C(C(=C1)O)N=C(C=C2)C(=O)O
Infrared Spectrum Authentic
Quantity 5g
Melting Point 213.0°C to 214.0°C
Color Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 189.17
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 8-Hydroxyquinoline-2-carboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.