Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
283158-18-9
C9H7NO4S·H2O
243.24
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
8-hydroxyquinoline-5-sulfonic acid hydrate,8-hydroxy-5-quinolinesulfonic acid monohydrate,8-hydroxyquinoline-5-sulfonic acid monohydrate,unii-n85kc14z44,8-hydroxy-5-quinolinesulfonic acid hydrate,5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1,8-hydroxyquinoline-5-sulfonic acid, hydrate,acmc-209pxa,dsstox_cid_24317,dsstox_rid_80150
2723649
8-hydroxyquinoline-5-sulfonic acid;hydrate
C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O
283158-18-9
C9H7NO4S·H2O
243.24
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
8-hydroxyquinoline-5-sulfonic acid hydrate,8-hydroxy-5-quinolinesulfonic acid monohydrate,8-hydroxyquinoline-5-sulfonic acid monohydrate,unii-n85kc14z44,8-hydroxy-5-quinolinesulfonic acid hydrate,5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1,8-hydroxyquinoline-5-sulfonic acid, hydrate,acmc-209pxa,dsstox_cid_24317,dsstox_rid_80150
2723649
8-hydroxyquinoline-5-sulfonic acid;hydrate
C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O
Melting Point >300.0°C
Color Yellow
Packaging Plastic bottle
Quantity 100g
Infrared Spectrum Authentic
Assay Percent Range 98%
Beilstein 22,407
Merck Index 14,4844
Solubility Information Solubility in water: freely soluble. Other solubilities: slightly soluble in most common organic solvents
Formula Weight 243.24
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 8-Hydroxyquinoline-5-sulfonic acid monohydrate
Show More Show Less

GHS H Statement
Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. Suspected of causing genetic defects.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Use personal protective equipment as required. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Obtain special instructions before use. Avoid breathing dust/fume/gas/mist/vapors/spray.

Warning