Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinoline Copper Salt, Approx. 98%, Spectrum™

Click to view available options
Quantity:
100 g
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10380-28-6
10380-28-6
Packaging Fiber Drum
Quantity 50 kg
CAS Max % 1
Percent Purity Approx. 98%

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.