Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinoline TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
148-24-3
C9H7NO
145.16
MCJGNVYPOGVAJF-UHFFFAOYSA-N
quinolin-8-ol
OC1=C2N=CC=CC2=CC=C1
148-24-3
C9H7NO
145.16
MCJGNVYPOGVAJF-UHFFFAOYSA-N
quinolin-8-ol
OC1=C2N=CC=CC2=CC=C1

H-163, 148-24-3

Spectrum™ 8-Hydroxyquinoline TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Packaging Poly Bottle
Quantity 500 mL
CAS Max % 1
UN Number UN1170