Alfa Aesar™ 8-Methoxyquinoline, 96%

Manufacturer: Alfa Aesar™ H5443003

Quantity 1g

 View more versions of this product

Catalog No. AAH5443003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 938-33-0
    Molecular Formula C10H9NO
    Formula Weight 159.19
    MDL Number MFCD00957068
    Chemical Name or Material 8-Methoxyquinoline

    Specifications

    Formula Weight 159.19
    CAS 938-33-0
    Melting Point 38° to 41°C
    Molecular Formula C10H9NO
    MDL Number MFCD00957068
    Quantity 1g
    Chemical Name or Material 8-Methoxyquinoline
    Assay 96%

    Description

    Applications
    8-Methoxyquinoline is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit