Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 8 mm Open Top Screw Thread Caps with Septa (8-425), 5.5 mm Hole Available on GSA/VA Contract for Federal Government customers only.

Fisherbrand™ 8 mm screw thread caps are chemically inert and suitable for most chromatography applications.

$53.60 - $65.60

Specifications

Closure Size 8 mm
Closure Type Screw
Color Black
Septum Red PTFE/White Silicone
No. per Pack 100
View More Specs

Products 2
Catalog Number Type Price Quantity & Availability  
Catalog Number Type Price Quantity & Availability  
03-391-46 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Autosampler Vial Screw Thread Closure
Pack of 100 for $53.60
Add to cart
 
13-622-211
View Documents
Autosampler Vial Screw Thread Closure with Flange
Pack of 100 for $65.60
Add to cart
 
Description

Description

  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Flanged caps are preferred for Shimadzu and Tosoh autosamplers
  • Pre-assembled caps and septa are convenient and minimize contamination from handling

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

8 mm
Black
100
Screw
Red PTFE/White Silicone
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit