Learn More
Cell Biolabs IncSupplier Diversity Partner 8-OHDG DNA DAMAGE ELISA TRL SZ  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA320T
The OxiSelect Oxidative DNA Damage ELISA Kit (Trial Size) provides rapid detection and quantitation of 8-OHdG in urine, plasma, serum, and DNA samples extracted from cells or tissues. 32 wells including standards and unknowns.
Catalog No. 50-673-502